siRNA Details
Gene Symbol: Gm29601 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076723.1 | - | 2 | 97 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB615092.1 | - | 2 | 97 | Chr. 1 - 96543763 - 96556703 [-] on Build GRCm38 |
