siRNA Details
Gene Symbol: 4933431K14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017009.1 | - | 1214 | |||
BB762387.1 | - | 404 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB020033.1 | - | 1 | 115 | Chr. 9 - 85750177 - 85750424 [+] on Build GRCm38 |
