siRNA Details
Gene Symbol: Gm33680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079646.1 | - | 113 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB633991.1 | - | 1 | 108 | Chr. 12 - 45466297 - 45492326 [+] on Build GRCm38 |
