siRNA Details
Gene Symbol: B230112J18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110475.1 | 3 | 330 | |||
GenBank mRNA | AK020970.1 | - | 3 | 331 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY727026.1 | - | 3 | 331 | Chr. 5 - 116312360 - 116318285 [+] on Build GRCm38 |
