siRNA Details
Gene Symbol: B430006N09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021015.1 | - | 1 | 320 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB749961.1 | - | 1 | 370 | Chr. 10 - 88415452 - 88415889 [+] on Build GRCm38 |
