siRNA Details
Gene Symbol: Gm5127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044840.1 | - | 4, 5 | 410 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB643050.1 | - | 4, 5 | 410 | Chr. X - 106583186 - 106708833 [+] on Build GRCm38 |
