siRNA Details
Gene Symbol: A930040O22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020947.1 | - | 1 | 593 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY742387.1 | - | 1 | 593 | Chr. 12 - 98831813 - 98832469 [+] on Build GRCm38 |
