siRNA Details
Gene Symbol: A430105D02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020766.1 | - | 1 | 938 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB750361.1 | - | 1 | 236 | Chr. 1 - 55009205 - 55009654 [-] on Build GRCm38 |
