siRNA Details
Gene Symbol: A930037H05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020940.1 | - | 1 | 306 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY742362.1 | - | 1 | 306 | Chr. 11 - 119490742 - 119491234 [-] on Build GRCm38 |
