siRNA Details
Gene Symbol: 4933432K03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102286.1 | 1 | 38 | |||
GenBank mRNA | AK005578.1 | - | 1 | 37 | ||
AK015302.1 | - | 39 | ||||
AK017025.1 | - | 37 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933432K03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102286.1 | 1 | 38 | |||
GenBank mRNA | AK005578.1 | - | 1 | 37 | ||
AK015302.1 | - | 39 | ||||
AK017025.1 | - | 37 |