siRNA Details
Gene Symbol: Gm13539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045340.1 | 3 | 200 | |||
GenBank mRNA | AK076911.1 | - | 3 | 202 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB615709.1 | - | 3 | 201 | Chr. 2 - 25784617 - 25787031 [+] on Build GRCm38 |
