siRNA Details
Gene Symbol: Gm31336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076914.1 | - | 4 | 671 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV270626.2 | - | 3 | 499 | Chr. 12 - 116742051 - 116744203 [-] on Build GRCm38 |
