siRNA Details
Gene Symbol: Gm31336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076914.1 | - | 2 | 118 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB615721.1 | - | 2 | 115 | Chr. 12 - 116742240 - 116757215 [-] on Build GRCm38 |
