siRNA Details
Gene Symbol: Gm5092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030245.1 | - | 1 | 16 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB614465.1 | - | 1 | 16 | Chr. 17 - 21091208 - 21098539 [+] on Build GRCm38 |
