siRNA Details
Gene Symbol: 1700125H03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038181.1 | 4 | 563 | |||
GenBank mRNA | AK015741.1 | - | 4 | 557 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715282.1 | - | 4 | 557 | Chr. 8 - 68368466 - 68375712 [+] on Build GRCm38 |
