siRNA Details
Gene Symbol: 1700125H03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038181.1 | 1, 2 | 78 | |||
NR_038189.1 | 1, 2 | 78 | ||||
GenBank mRNA | AK007280.1 | - | 1, 2 | 72 | ||
AK015741.1 | - | 1, 2 | 72 | |||
AK131689.1 | - | 1, 2 | 79 |
