siRNA Details
Gene Symbol: Gm26604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048019.1 | - | 90 | |||
BC098190.1 | - | 1 | 56 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB648634.1 | - | 1 | 87 | Chr. 7 - 29071606 - 29080054 [+] on Build GRCm38 |
