siRNA Details
Gene Symbol: Gm9926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040528.1 | 3 | 701 | |||
GenBank mRNA | AK033454.1 | - | 3 | 665 | ||
AK136461.1 | - | 3 | 702 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB624834.1 | - | 3 | 662 | Chr. 18 - 66504183 - 66507575 [+] on Build GRCm38 |
