siRNA Details
Gene Symbol: 4930405J17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045350.1 | 3 | 563 | |||
GenBank mRNA | AK015099.1 | - | 3 | 549 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU937526.1 | - | 3 | 566 | Chr. 10 - 20199219 - 20201854 [-] on Build GRCm38 |
