siRNA Details
Gene Symbol: 4930405A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015090.1 | - | 1 | 365 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714619.1 | - | 1 | 366 | Chr. 11 - 103809392 - 103809998 [-] on Build GRCm38 |
