siRNA Details
Gene Symbol: Gm29230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132084.1 | - | 2 | 241 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712925.1 | - | 2 | 237 | Chr. 3 - 51058908 - 51074852 [-] on Build GRCm38 |
