siRNA Details
Gene Symbol: 6330565B04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018240.1 | - | 1 | 464 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY718691.1 | - | 1 | 464 | Chr. 1 - 28570488 - 28571431 [-] on Build GRCm38 |
