siRNA Details
Gene Symbol: 4930547E08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040514.1 | 3, 4 | 338 | |||
GenBank mRNA | AK019749.1 | - | 3, 4 | 337 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715652.1 | - | 3, 4 | 334 | Chr. 2 - 103804452 - 103810619 [-] on Build GRCm38 |
