siRNA Details
Gene Symbol: 4930546O09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019747.1 | - | 1 | 201 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715650.1 | - | 1 | 201 | Chr. 16 - 46021945 - 46027375 [-] on Build GRCm38 |
