siRNA Details
Gene Symbol: 4930545L08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019741.1 | - | 1 | 293 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715639.1 | - | 1 | 293 | Chr. 9 - 111912311 - 111918666 [-] on Build GRCm38 |
