siRNA Details
Gene Symbol: 1810012K08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007459.1 | - | 1 | 82 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707528.1 | - | 1 | 81 | Chr. 2 - 26138546 - 26139543 [+] on Build GRCm38 |
