siRNA Details
Gene Symbol: C79798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138624.1 | - | 2 | 1184 | ||
BG079793.2 | - | 1 | 203 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C79798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138624.1 | - | 2 | 1184 | ||
BG079793.2 | - | 1 | 203 |