siRNA Details
Gene Symbol: 9330178D15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040553.1 | 1 | 21 | |||
GenBank mRNA | AK034327.1 | - | 1 | 22 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB626353.1 | - | 1 | 19 | Chr. 3 - 155976356 - 156005802 [+] on Build GRCm38 |
