siRNA Details
Gene Symbol: 7530416G11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK078701.1 | - | 4, 5 | 523 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB624027.1 | - | 4, 5 | 518 | Chr. 15 - 85493239 - 85503223 [-] on Build GRCm38 |
