siRNA Details
Gene Symbol: 9030411M13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018502.1 | - | 1 | 1523 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ228391.1 | - | 1 | 101 | Chr. 10 - 125221650 - 125222361 [-] on Build GRCm38 |
