siRNA Details
Gene Symbol: 9530027D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020577.1 | - | 1 | 576 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB485726.2 | - | 1 | 507 | Chr. 13 - 107852002 - 107852672 [-] on Build GRCm38 |
