siRNA Details
Gene Symbol: 9530025L08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020572.1 | - | 1 | 437 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY722053.1 | - | 1 | 437 | Chr. 19 - 30165114 - 30165769 [-] on Build GRCm38 |
