siRNA Details
Gene Symbol: 1700025C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033448.1 | 2 | 249 | |||
GenBank mRNA | AK006325.1 | - | 2 | 249 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706397.1 | - | 2 | 249 | Chr. 2 - 165078721 - 165090750 [-] on Build GRCm38 |
