siRNA Details
Gene Symbol: Gm11725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090182.1 | - | 1 | 41 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB869472.1 | - | 1 | 41 | Chr. 11 - 117871250 - 117872149 [+] on Build GRCm38 |
