siRNA Details
Gene Symbol: 4930535I16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050163.1 | - | 1 | 1003 | ||
AK085801.1 | - | 1 | 1027 | |||
BB667541.1 | - | 1 | 407 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF723499.1 | - | 1 | 114 | Chr. 4 - 123915770 - 123916385 [-] on Build GRCm38 |
