siRNA Details
Gene Symbol: 1700020B03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006142.1 | - | 1 | 367 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706248.1 | - | 1 | 367 | Chr. 16 - 46183152 - 46183604 [-] on Build GRCm38 |
