siRNA Details
Gene Symbol: Gm4632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC027670.1 | - | 3 | 515 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CO807057.1 | - | 3 | 489 | Chr. 14 - 79178893 - 79247289 [-] on Build GRCm38 |
