siRNA Details
Gene Symbol: Gm4632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040428.1 | - | 1 | 177 | ||
BC027670.1 | - | 1 | 88 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA856347.1 | - | 1 | 87 | Chr. 14 - 79179020 - 79199190 [-] on Build GRCm38 |
