siRNA Details
Gene Symbol: Dreh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105051.1 | 2 | 597 | |||
GenBank mRNA | AK050349.1 | - | 2 | 598 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI451793.1 | - | 2 | 433 | Chr. 17 - 64767275 - 64767904 [+] on Build GRCm38 |
