siRNA Details
Gene Symbol: 2310015A10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009348.1 | - | 1 | 26 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX215131.1 | - | 1 | 26 | Chr. 12 - 80124983 - 80132596 [-] on Build GRCm38 |
