siRNA Details
Gene Symbol: NDUFA6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034118.2 | 5 | 1298 | |||
GenBank mRNA | AK123630.1 | - | 5 | 1300 |
Gene Symbol: LOC102723722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029174.1 | XP_016884663.1 | 4 | 521 |
