siRNA Details
Gene Symbol: C20orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242671.2 | NP_001229600.1 | 3 | 374 | ||
NM_001302818.1 | NP_001289747.1 | 3 | 350 | |||
GenBank mRNA | BX112808.1 | - | 3 | 324 | ||
BX334499.2 | - | 3 | 341 | |||
HY016900.1 | - | 3 | 308 | |||
HY050710.1 | - | 3 | 375 |
