siRNA Details
Gene Symbol: THEG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205273.2 | NP_001192202.1 | 2 | 416 | ||
NM_001278575.1 | NP_001265504.1 | 3 | 826 | |||
NM_001278576.1 | NP_001265505.1 | 4 | 950 | |||
NM_001278577.1 | NP_001265506.1 | 4 | 961 | |||
NM_001278578.1 | NP_001265507.1 | 3 | 819 | |||
XM_011526302.2 | XP_011524604.1 | 3 | 2605 | |||
XM_011526303.2 | XP_011524605.1 | 3 | 1430 | |||
XM_017026126.1 | XP_016881615.1 | 3 | 2941 | |||
XM_017026127.1 | XP_016881616.1 | 3 | 2400 | |||
XM_017026128.1 | XP_016881617.1 | 3 | 2598 | |||
XM_017026129.1 | XP_016881618.1 | 3 | 1437 | |||
XM_017026130.1 | XP_016881619.1 | 3 | 1162 | |||
XM_017026131.1 | XP_016881620.1 | 4 | 768 | |||
XM_017026132.1 | XP_016881621.1 | 3 | 961 | |||
XM_017026133.1 | XP_016881622.1 | 2 | 629 | |||
GenBank mRNA | BC043286.1 | - | 4 | 889 | ||
FJ211597.1 | - | 2 | 416 |
