siRNA Details
Gene Symbol: LINC00659 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046224.1 | 3 | 402 | |||
NR_046225.1 | 3 | 268 | ||||
GenBank mRNA | BM850706.1 | - | 3 | 287 | ||
DA633805.1 | - | 3 | 467 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00659 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046224.1 | 3 | 402 | |||
NR_046225.1 | 3 | 268 | ||||
GenBank mRNA | BM850706.1 | - | 3 | 287 | ||
DA633805.1 | - | 3 | 467 |