siRNA Details
Gene Symbol: HDAC11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046690.1 | 1 | 352 | |||
GenBank mRNA | BX104155.1 | - | 1 | 352 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HDAC11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046690.1 | 1 | 352 | |||
GenBank mRNA | BX104155.1 | - | 1 | 352 |