siRNA Details
Gene Symbol: LOC101928185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135572.1 | 2 | 1015 | |||
GenBank mRNA | BC031872.1 | - | 2 | 1001 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135572.1 | 2 | 1015 | |||
GenBank mRNA | BC031872.1 | - | 2 | 1001 |