siRNA Details
Gene Symbol: LOC101926948 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135130.1 | 1, 2 | 173 | |||
GenBank mRNA | BC042185.1 | - | 1, 2 | 161 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI838016.1 | - | 1, 2 | 161 | Chr. 9 - 35646270 - 35647093 [+] on Build GRCh38 |
