siRNA Details
Gene Symbol: LOC101926963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110054.1 | 2 | 545 | |||
NR_110055.1 | 2 | 311 | ||||
GenBank mRNA | BY993667.1 | - | 2 | 311 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101926963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110054.1 | 2 | 545 | |||
NR_110055.1 | 2 | 311 | ||||
GenBank mRNA | BY993667.1 | - | 2 | 311 |