siRNA Details
Gene Symbol: LOC101926963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110054.1 | 1, 2 | 462 | |||
NR_110055.1 | 1, 2 | 228 | ||||
GenBank mRNA | BX114951.1 | - | 1, 2 | 462 | ||
BY993667.1 | - | 1, 2 | 228 |
