siRNA Details
Gene Symbol: DLG1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038289.1 | 3 | 588 | |||
GenBank mRNA | BC039329.1 | - | 3 | 583 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI460168.1 | - | 3 | 587 | Chr. 3 - 197298247 - 197302698 [+] on Build GRCh38 |
